EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Preparation, assay, and microinjection of fluorescently labeled cytoskeletal proteins: actin, alpha-actinin, and vinculin.
PMID 3102901
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
86186
Sulforhodamine B acid chloride, suitable for fluorescence, technical
C27H29ClN2O6S2