EMAIL THIS PAGE TO A FRIEND
Biochimie
One step purification of Escherichia coli beta-glucuronidase.
PMID 3105604
Abstract
beta-glucuronidase was purified by affinity chromatography on thiophenyl-glucuronide coupled to Sepharose. The enzyme was more than 95% pure. This enzyme is a tetramer composed of identical 74 kDa monomers. The amino-terminal sequence determined was: NH2-Met-Leu-Arg-Pro-Val.