EMAIL THIS PAGE TO A FRIEND
Developments in biological standardization
A rapid and sensitive method for the detection of mycoplasmas in infected cell cultures using 6-methyl purine deoxyriboside.
PMID 3108056
Abstract
6MPDR causes the death of cells lightly infected with mycoplasmas. This is brought about by the action of the mycoplasmal enzyme adenosine phosphorylase which converts 6MPDR to highly toxic metabolites. If the medium conditions are adjusted to favour the growth of mycoplasmas by the addition of pig serum to the medium, infections as low as 1 mycoplasma per 200,000 cells can be detected within 7 days. This finding enables mycoplasma tests to be carried out rapidly and reliably by untrained personnel.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M6502 6-Methylpurine, ≥99%
C6H6N4