EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
The structure of inducing factors for virginiamycin production in Streptomyces virginiae.
PMID 3108224
Abstract
Virginiamycin inducing factors (inducing material or inducing factor) of Streptomyces virginiae were isolated from the culture broth of this microbe and separated into three closely related compounds. They were named virginiae butanolides A, B and C and their structures were determined as 2-(1'-hydroxy-5'-methylhexyl)-3-(hydroxymethyl)butanolide (6), 2-(1'-hydroxy-4'-methylhexyl)-3-(hydroxymethyl)butanolide (7) and 2-(1'-hydroxyhexyl)-3-(hydroxymethyl)butanolide (8), respectively. Part of their stereochemistry was also determined. Racemic virginiae butanolide C was synthesized to confirm their structures.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00045
Virginiaebutanolide C, ≥90% (LC/MS-ELSD)
C11H20O4