EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Novel phenylalanine dehydrogenases from Sporosarcina ureae and Bacillus sphaericus. Purification and characterization.
PMID 3112142
Abstract
NAD+-dependent phenylalanine dehydrogenases were purified 1,500- and 1,600-fold, and crystallized from Sporosarcina ureae SCRC-R04 and Bacillus sphaericus SCRC-R79a, respectively. The purified enzymes were homogeneous as judged by disc gel electrophoresis. The enzyme from S. ureae has a molecular weight of 305,000, while that of B. sphaericus has a molecular weight of 340,000. Each is probably composed of eight subunits identical in molecular weight. The S. ureae enzyme showed a high substrate specificity in the oxidative deamination reaction acting on L-phenylalanine, while that of B. sphaericus acted on L-phenylalanine and L-tyrosine. The enzymes had lower substrate specificities in the reductive amination reaction acting on alpha-keto acids. The Sporosarcina enzyme acted on phenylpyruvate, alpha-ketocaproate, alpha-keto-gamma-methylthiobutyrate and rho-hydroxyphenylpyruvate. The Bacillus enzyme acted on rho-hydroxyphenylpyruvate, phenylpyruvate, and alpha-keto-gamma-methylthiobutyrate. The enzyme from B. sphaericus catalyzes The enzyme from B. sphaericus catalyzes the transfer of pro-S (B) hydrogen from NADH.
Related Materials