EMAIL THIS PAGE TO A FRIEND
Xenobiotica; the fate of foreign compounds in biological systems
The measurement of ethylenethiourea and ethyleneurea in the rat and common marmoset Callithrix jacchus after zineb (zinc ethylenebisdithiocarbamate) dosing.
PMID 3114968
Abstract
The metabolism of the fungicide zineb, (zinc ethylenebisdithiocarbamate), has been studied in the rat and the marmoset. 2. It was found in both species that a relatively large proportion (21-22%) of the original zineb administered was detectable in the excreta as ethylenethiourea (ETU) a known mutagen, teratogen and carcinogen. 3. A further proportion (2-5%) was determined to be ethyleneurea which is a metabolite of ETU. 4. Results of comparative experiments in marmosets revealed that ETU was photolabile in the presence of excreta, thus showing the importance of conducting the studies in the dark.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31534
2-Imidazolidone, PESTANAL®, analytical standard
C3H6N2O
I601
2-Imidazolidone, 96%
C3H6N2O