EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Active-site titration of serine proteinases acting selectively on cationic substrates by N-alpha-carbobenzoxy-L-arginine p-nitrophenyl ester and N-alpha-carbobenzoxy-L-lysine p-nitrophenyl ester; determination of active enzyme concentration.
PMID 3115299
Abstract
A titration method for determination of trypsin-like serine proteinase concentration has been developed by using ZArgONp and ZLysONp, two specific chromogenic amino-acid derivatives which show the characteristics of optimal active-site titrants. Active proteinase concentration has been estimated from the effect of titrant concentration on the amplitude, at time zero, of the time-course for the instantaneous release of p-nitrophenol, preceding the steady-state reaction (burst phase).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3637
Z-L-Lys-ONp hydrochloride
C20H23N3O6 · HCl
96895
Z-Lys-ONp hydrochloride, ≥98.0% (AT)
C20H23N3O6 · HCl