EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Cloning, sequencing, and expression of cDNA for human beta-galactosidase.
PMID 3143362
Abstract
We cloned and sequenced the full-length cDNA for human placental beta-galactosidase. The 2379-nucleotide sequence contains 2031 nucleotides which encode a protein of 677 amino acids. The amino acid sequence includes a putative signal sequence of 23 amino acids and 7 potential asparagine-linked glycosylation sites. The cDNA in the expression vector pSVL was used to transfect COS cells. Expression of the cDNA in transfected COS cells produced immunoprecipitable proteins and led to an increase in beta-galactosidase activity.