EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Toxic effects of mercuric chloride, methylmercuric chloride, and emisan 6 (an organic mercurial fungicide) on ovarian recrudescence in the catfish Clarias batrachus (L.).
PMID 3148347