EMAIL THIS PAGE TO A FRIEND
Acta poloniae pharmaceutica
[Studies of pyrazine derivatives. XXV. Synthesis and the tuberculostatic activity of the products of reaction of 5-(6-methoxy- and 5-(6 morpholinopyrazinyl)-1,3,4-oxadiazol-2-thione with amines].
PMID 3150245
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M72609
N-Methylpiperidine, 99%
C6H13N