EMAIL THIS PAGE TO A FRIEND
Indian journal of experimental biology
Inhibition of testicular 3 beta-hydroxy-delta 5-steroid dehydrogenase (3 beta-HSD) activity in catfish Clarias batrachus (L.) by mercurials.
PMID 3150370