EMAIL THIS PAGE TO A FRIEND
Annales Universitatis Mariae Curie-Sklodowska. Sectio D: Medicina
Topochemical examinations of the lungs of the rat after combined administration of serpasil and deslanatoside. I. Histochemical examinations.
PMID 3154882
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0600000 Deslanoside, European Pharmacopoeia (EP) Reference Standard
C47H74O19