EMAIL THIS PAGE TO A FRIEND
Indian journal of ophthalmology
A comparative study between intra-vitreal and sub-conjunctival antibiotic in bacterial endophthalmitis.
PMID 3155135
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D1928
Fenaminosulf, ≥98%
C8H10N3NaO3S
45482
Fenaminosulf, PESTANAL®, analytical standard
C8H10N3NaO3S