EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
An improved method for the preparation of derivatives of reducing oligosaccharide with 2-(4-aminophenyl)ethylamine.
PMID 3167886
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
123056
4-(2-Aminoethyl)aniline, 97%
C8H12N2