EMAIL THIS PAGE TO A FRIEND
IARC scientific publications
Determination of methylhippuric acids in urine by gas chromatography.
PMID 3182055
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
328006
2-Methylhippuric acid, 98%
C10H11NO3
328014
3-Methylhippuric acid, 98%
C10H11NO3
328022
4-Methylhippuric acid, 98%
C10H11NO3