EMAIL THIS PAGE TO A FRIEND
Hepatology (Baltimore, Md.)
Do branched chain keto amino acids regulate their own metabolism?
PMID 3192188
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
K6625
2-Ketohexanoic acid sodium salt, 97.0-103.0%
C6H9NaO3