EMAIL THIS PAGE TO A FRIEND
Nihon juigaku zasshi. The Japanese journal of veterinary science
Therapeutic effect of isoprothiolane on bovine fat necrosis.
PMID 3199611
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
76547
Isoprothiolane, analytical standard
C12H18O4S2