EMAIL THIS PAGE TO A FRIEND
Archives of disease in childhood
Mercury as a health hazard.
PMID 3202653
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
10005
Mercury(II) amidochloride, ≥97.0%, powder
H2ClHgN