EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Interconversion model analysis and estimation of available fractions for 4-acetaminobenzaldehyde and its reversible metabolite, 4-acetaminobenzylalcohol, in the rat using the multi-lines fitting technique.
PMID 3203428
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1800
4-Acetamidobenzaldehyde, technical grade
C9H9NO2