EMAIL THIS PAGE TO A FRIEND
Journal of the South African Veterinary Association
Propethampos, cyromazine and blowfly strike in sheep.
PMID 3210212
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34371
Propetamphos, PESTANAL®, analytical standard
C10H20NO4PS