EMAIL THIS PAGE TO A FRIEND
Die Pharmazie
[HPLC determination of benzoylmetronidazole and metronidazole in biological materials].
PMID 3212032
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1851000 Metronidazole benzoate, European Pharmacopoeia (EP) Reference Standard
C13H13N3O4