EMAIL THIS PAGE TO A FRIEND
Journal de toxicologie clinique et experimentale
[Experimental toxicity of indium formate and indium formate- dibutyltin oxide mixture (90-10) in the rat].
PMID 3216342
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
183083
Dibutyltin(IV) oxide, 98%
C8H18OSn
34980
Dibutyltin(IV) oxide, purum
C8H18OSn