EMAIL THIS PAGE TO A FRIEND
The Veterinary record
Treatment of Dicrocoelium dendriticum with a combination of thiophanate and brotianide.
PMID 3218049
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34078
Oxyclozanide, VETRANAL, analytical standard
C13H6Cl5NO3