EMAIL THIS PAGE TO A FRIEND
Il Farmaco; edizione pratica
Binding of theophylline, ethophylline and diprophylline to human serum albumin.
PMID 3234537
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H9006
7-(β-Hydroxyethyl)theophylline, solid
C9H12N4O3