EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Formation of diastereomeric derivatives of 2-arylpropionic acids using L-leucinamide: lack of generality.
PMID 3235535
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
286427
L-Leucinamide hydrochloride, 99%
C6H14N2O · HCl