EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Measurement of carprofen enantiomer concentrations in plasma and urine using L-leucinamide as the chiral coupling component.
PMID 3235564
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L8250
DL-Leucinamide hydrochloride
C6H14N2O · HCl
286427
L-Leucinamide hydrochloride, 99%
C6H14N2O · HCl