EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Trans-4-methylcyclohexylamine, a potent new inhibitor of spermidine synthase.
PMID 3240528
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
177466
4-Methylcyclohexylamine, mixture of cis and trans, 97%
C7H15N