EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Thin-layer chromatography of flucythrinate residues.
PMID 3241026
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33496
Flucythrinate, PESTANAL®, analytical standard
C26H23F2NO4