EMAIL THIS PAGE TO A FRIEND
Il Farmaco; edizione pratica
Determination of optical purity by high-performance liquid chromatography of compounds of pharmaceutical interest.
PMID 3243351
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A43804
2-Amino-1-butanol, 97%
C4H11NO