EMAIL THIS PAGE TO A FRIEND
Gigiena truda i professional'nye zabolevaniia
[Hygienic standardization of monochloracetic acid chloranhydride in the air of the work area].
PMID 3243499
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
22880
Chloroacetyl chloride, purum, ≥99.0% (GC)
C2H2Cl2O
104493
Chloroacetyl chloride, 98%
C2H2Cl2O