EMAIL THIS PAGE TO A FRIEND
Gigiena truda i professional'nye zabolevaniia
[A case of acute methylchlorformiate poisoning].
PMID 3243500
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M35304
Methyl chloroformate, 99%
C2H3ClO2