EMAIL THIS PAGE TO A FRIEND
Eisei Shikenjo hokoku. Bulletin of National Institute of Hygienic Sciences
[Hepatic ornithine decarboxylase activity of rats induced by administration of 2-nitro-p-cresol and sodium deoxycholate].
PMID 3250663