EMAIL THIS PAGE TO A FRIEND
Arzneimittel-Forschung
[The binding of oxicam derivatives to human serum albumin].
PMID 3264167
Abstract
The binding characteristics of several oxicam derivatives (tenoxicam, 4-hydroxy-2-methyl-N-phenyl-2H-1,2-benzothiazixine-3- carboxamide-1,1-dioxide (CP 14,304), 4-hydroxy-2-methyl-N-2-(3-methyl)-pyridyl-2H-1,2-benzothiazixine-3 -carboxamide- 1,1-dioxide (CP 16,460), piroxicam, meloxicam [corrected], isoxicam, 5-hydroxy-piroxicam) to 2% and 4% human serum albumin (HSA) were determined using a modified ultrafiltration process. The binding properties to HSA were characterized by determining the overall binding constant, the apparent binding constant, the slope, the free reaction energy, and the unbound portion of the drug. The following results were obtained: 1. These oxicam derivatives show a high affinity to HSA. The unbound fraction amounts to 1-3%. 2. The affinity of the compounds to HSA decreases in the order mentioned above. 3. Doubling of the HSA concentration reduces the unbound fraction, to the half, with piroxicam being the only exception.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32122
5′-Hydroxypiroxicam, VETRANAL, analytical standard
C15H13N3O5S
I1762
Isoxicam, analytical standard
C14H13N3O5S