EMAIL THIS PAGE TO A FRIEND
Oral microbiology and immunology
Activity of metronidazole and its hydroxy metabolite against clinical isolates of Actinobacillus actinomycetemcomitans.
PMID 3268747
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34007
Metronidazole-OH, VETRANAL, analytical standard
C6H9N3O4