EMAIL THIS PAGE TO A FRIEND
Prostaglandins
The biology and pharmacology of PGD2.
PMID 3283848
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5172
Prostaglandin D2, ≥95%, synthetic
C20H32O5