EMAIL THIS PAGE TO A FRIEND
Journal de toxicologie clinique et experimentale
Hydroxocobalamin in chronic cyanide poisoning.
PMID 3286861
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H1428000 Hydroxocobalamin, European Pharmacopoeia (EP) Reference Standard
C62H89CoN13O15P