EMAIL THIS PAGE TO A FRIEND
Progress in lipid research
Platelet activating factor; chemical and biochemical characteristics.
PMID 3313430
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P4904
β-Acetyl-γ-O-hexadecyl-L-α-phosphatidylcholine hydrate, ≥98%
C26H54NO7P · xH2O