EMAIL THIS PAGE TO A FRIEND
Annales de biologie clinique
[5-Fluorocytosine and combinations].
PMID 3326438
Abstract
The antimycotic and pharmacological properties of 5-fluorocytosine are presented. Especially the aspect of good gastrointestinal absorption, elimination by the kidney, unwanted side effects and appearance of primary and secondary resistance are discussed. The indication of 5 FC monotherapy or combination therapy is shown in various manifestations of deep candidiasis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F0175000
Flucytosine, European Pharmacopoeia (EP) Reference Standard
C4H4FN3O