EMAIL THIS PAGE TO A FRIEND
Neuroscience letters
Presynaptic M1 muscarinic receptor modulates spontaneous release of acetylcholine from rat basal forebrain slices.
PMID 3340327
Abstract
Spontaneous release of acetylcholine (ACh) from rat basal forebrain slices in the presence of cholinesterase inhibitor was directly determined using a specific radioimmunoassay for ACh. The release was calcium dependent. A consistent amount of ACh release was observed throughout the experiment. Atropine (10(-8) to 10(-5) M) and pirenzepine (10(-7) to 10(-5) M) enhanced spontaneous ACh release. These findings indicate the presence of an M1 muscarinic autoreceptor that modulates spontaneous release of ACh in the rat basal forebrain.