EMAIL THIS PAGE TO A FRIEND
Toxicology
Response of blood and brain cholinesterase to dermal exposure of bromophos in the rat.
PMID 3341046
Abstract
Response of cholinesterase to dermal exposure of acute, single and multiple doses of Bromophos in the female rat has been studied. Dose-response studies (50-4000 mg/kg body weight, 24 h exposure) showed that plasma cholinesterase was most sensitive to inhibition in vivo, followed by the brain and erythrocyte acetylcholinesterase. The ID50 values for the in vivo cholinesterase inhibition were 10.1, 576.1 and 1938.0 mg/kg body weight for the plasma, brain and erythrocytes, respectively. In time-course studies after a single sublethal dose of 1000 mg/kg body weight (24 h) of Bromophos, the serum and brain cholinesterase were rapidly inhibited reaching a maximum at 16 h. Recovery was complete in the case of serum at 14 days post-exposure, whereas the brain enzyme was not fully recovered at 21 days. In a subacute study, daily dermal application of 50 mg/kg body weight of Bromophos for 5 and 10 days, resulted in high inhibition of the serum cholinesterase and brain acetylcholinesterase, the former being more marked which was reversible after 10 or 15 days of post-exposure period. Very low levels of dermal exposure of Bromophos (10-50 mg/kg body weight) for 17 days caused pronounced depression of serum cholinesterase which completely recovered in 15 days after cessation of exposure suggesting that the serum cholinesterase could serve as the most sensitive diagnostic indicator of Bromophos exposure.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33400
Bromophos-methyl, PESTANAL®, analytical standard
C8H8BrCl2O3PS