EMAIL THIS PAGE TO A FRIEND
Journal of steroid biochemistry
Aromatization of testosterone and 19-nortestosterone by a single enzyme from equine testicular microsomes. Differences from human placental aromatase.
PMID 3347045
Abstract
A single enzyme in the stallion testis was able to aromatize both testosterone and nortestosterone. This enzyme had a much lower affinity for nortestosterone than for testosterone. In contrast to human placental estrogen synthetase, this enzyme aromatized testosterone and 19-nortestosterone with similar efficiency. The differences observed (effects of monovalent cations, inhibition of androstenedione aromatization by testosterone and 19-nortestosterone and, above all, rate of norandrogen aromatization) suggest that the aromatase in the horse testis is not the same as that in the human placenta.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7252
19-Nortestosterone
C18H26O2