EMAIL THIS PAGE TO A FRIEND
Gigiena truda i professional'nye zabolevaniia
[Experimental data for the substantiation of the maximum allowable concentration of meta-aminophenol in the air of work areas].
PMID 3356351