EMAIL THIS PAGE TO A FRIEND
Analytical chemistry
Speciation of methyl- and butyltin compounds and inorganic tin in oysters by hydride generation atomic absorption spectrometry.
PMID 3358487
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
201057
Butyltin trichloride, 95%
C4H9Cl3Sn