EMAIL THIS PAGE TO A FRIEND
Journal of toxicology and environmental health
Metabolism of 1-, 3-, and 6-nitrobenzo[a]pyrene by intestinal microflora.
PMID 3361619
Abstract
The compounds 1-, 3-, and 6-nitrobenzo[a]pyrene (nitro-BaP) are environmental pollutants and have been shown to be potent bacterial mutagens. The anaerobic metabolism of these isomeric nitro-BaPs was investigated by the incubation of rat intestinal microflora with each isomer for 48 h. Aliquots were removed at several time intervals, extracted, fractionated by high-pressure liquid chromatography (HPLC), and the radioactivity determined. Metabolites were identified by comparison of their chromatographic, ultraviolet-visible absorption, and mass spectral properties with those of authentic standards. The order of the extent of nitroreduction for these isomers was 3-nitro-BaP greater than 6-nitro-BaP greater than 1-nitro-BaP. After 48 h of exposure, 84% of the added 3-nitro-BaP was present as 3-amino-BaP, 51% of the 6-nitro-BaP was metabolized to 6-amino-BaP, and 1-nitro-BaP was reduced to 1-amino-BaP (13%) and 1-nitro-BaP (4%). The order of the extent of microbial nitroreduction for these nitro-BaP isomers is different from the predictions based on electronic and steric hindrance effects. These results suggest that intestinal microflora nitroreductases exhibit a markedly high degree of substrate specificity toward nitro-BaPs that affects the extent of nitroreduction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
BCR311
6-Nitrobenzo[a]pyrene, BCR® certified Reference Material
C20H11NO2