EMAIL THIS PAGE TO A FRIEND
European journal of biochemistry
Characterization of the isolated transferase subunit of citrate lyase as a CoA-Transferase. Evidence against a covalent enzyme-substrate intermediate.
PMID 336371