EMAIL THIS PAGE TO A FRIEND
Archives of microbiology
Bacterial metabolism of side chain fluorinated aromatics: cometabolism of 3-trifluoromethyl(TFM)-benzoate by Pseudomonas putida (arvilla) mt-2 and Rhodococcus rubropertinctus N657.
PMID 3365096
Abstract
The TOL plasmid-encoded enzymes of the methylbenzoate pathway in Pseudomonas putida mt-2 cometabolized 3-trifluoromethyl (TFM)-benzoate. Two products, 3-TFM-1,2-dihydroxy-2-hydrobenzoate (3-TFM-DHB) and 2-hydroxy-6-oxo-7,7,7-trifluoro-hepta-2,4-dienoate (7-TFHOD) were identified chemically and by spectroscopic properties. TFM-substituted analogues of the metabolites of the methylbenzoate pathway were generally converted at drastically reduced rates. The catechol-2,3-dioxygenase from Pseudomonas putida showed moderate turnover rates with 3-TFM-catechol. The catechol-1,2-dioxygenase of Rhodococcus rubropertinctus N657 was totally inhibited by 3-TFM-catechol and did not cleave this substrate. Hammett-type analysis showed the catechol-1,2-dioxygenase reaction to be strongly dependent on the electronic nature of the substituents. Electronegative substituents strongly inhibited catechol cleavage. The catechol-2,3-dioxygenase reaction, however, was only moderately sensitive to electronegative substituents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
188344
3-(Trifluoromethyl)benzoic acid, 99%
C8H5F3O2