EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Allergic contact dermatitis from an ostomy bag.
PMID 3365958
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
533203
2-(2-Hydroxy-5-methylphenyl)benzotriazole, 97%
C13H11N3O