EMAIL THIS PAGE TO A FRIEND
International journal of peptide and protein research
Heterochiral N-formyl methionyl peptides. Molecular conformation and crystal structures of N-f-L-Met-D-Phe and N-f-L-Met-D-Phe-OtBu.
PMID 3366550