EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Spontaneous recovery of cholinesterases after organophosphate intoxication: effect of environmental temperature.
PMID 3370329
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P2880
Propionylthiocholine iodide, ≥98%
C8H18INOS