EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Photometric determination of 3-chloroacetyl indole in the air].
PMID 3371676
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0482
3CAI, ≥95% (HPLC)
C10H8ClNO